Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046241.1 | NP_001039706.1 | 4 - 5 | 336 | 115 | |
| GenBank mRNA | BC105329.1 | - | 4 - 5 | 336 | 115 | |
More Information
Set Membership: |
|

RBX1
Genomic Map