Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046248.2 | NP_001039713.1 | 1 - 2 | 138 | 52 | |
| GenBank mRNA | BC114023.1 | - | 1 - 2 | 140 | 52 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114023.1, NM_001046248.2 transcripts.
Set Membership: |
|

PLPPR2
Genomic Map