Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046248.2 | NP_001039713.1 | 4 - 5 | 523 | 118 | |
| GenBank mRNA | BC114023.1 | - | 4 - 5 | 525 | 118 | |
More Information
Set Membership: |
|

PLPPR2
Genomic Map