Assay Details
| Gene Symbol: PKNOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046350.1 | NP_001039815.1 | 11 - 12 | 1364 | 54 | |
| GenBank mRNA | BC113256.1 | - | 11 - 12 | 1364 | 54 | |
| BT026210.1 | - | 9 - 10 | 1083 | 54 | ||
More Information
Set Membership: |
|

PKNOX1
Genomic Map