Assay Details
| Gene Symbol: TENT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046361.1 | NP_001039826.1 | 14 - 15 | 1719 | 89 | |
| GenBank mRNA | BC113319.1 | - | 14 - 15 | 1719 | 89 | |
More Information
Set Membership: |
|

TENT2
Genomic Map