Assay Details
| Gene Symbol: CALHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046420.1 | NP_001039885.1 | 2 - 3 | 959 | 62 | |
| GenBank mRNA | BC113290.1 | - | 2 - 3 | 959 | 62 | |
More Information
Set Membership: |
|

CALHM2
Genomic Map