Assay Details
| Gene Symbol: SOX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046429.2 | NP_001039894.1 | 1 - 2 | 1314 | 77 | |
| GenBank mRNA | BC114054.1 | - | 1 - 2 | 1315 | 77 | |
More Information
Set Membership: |
|

SOX30
Genomic Map