Assay Details
| Gene Symbol: PPIL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046537.2 | NP_001040002.1 | 6 - 7 | 681 | 97 | |
| GenBank mRNA | BC114789.1 | - | 6 - 7 | 683 | 97 | |
More Information
Set Membership: |
|

PPIL6
Genomic Map