Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046573.1 | NP_001040038.1 | 1 - 2 | 120 | 108 | |
| GenBank mRNA | BC113222.1 | - | 1 - 2 | 120 | 108 | |
More Information
Set Membership: |
|

SNAPC5
Genomic Map