Assay Details
| Gene Symbol: CCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075147.2 | NP_001068615.1 | 1 - 2 | 152 | 87 | |
| GenBank mRNA | BC118471.1 | - | 1 - 2 | 153 | 87 | |
More Information
Set Membership: |
|

CCL4
Genomic Map