Assay Details
| Gene Symbol: TPMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075531.1 | NP_001068999.1 | 6 - 7 | 585 | 69 | |
| GenBank mRNA | BC118238.1 | - | 6 - 7 | 585 | 69 | |
More Information
Set Membership: |
|

TPMT
Genomic Map