Assay Details
| Gene Symbol: APOO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075639.1 | NP_001069107.1 | 5 - 6 | 404 | 88 | |
| GenBank mRNA | BC118339.1 | - | 5 - 6 | 404 | 88 | |
More Information
Set Membership: |
|

APOO
Genomic Map