Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075680.2 | NP_001069148.1 | 8 - 9 | 1190 | 73 | |
| GenBank mRNA | BC114044.1 | - | 8 - 9 | 1192 | 73 | |
More Information
Set Membership: |
|

AP3M1
Genomic Map