Assay Details
| Gene Symbol: GEMIN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075731.2 | NP_001069199.1 | 3 - 4 | 607 | 108 | |
| GenBank mRNA | BC116157.1 | - | 3 - 4 | 609 | 108 | |
More Information
Set Membership: |
|

GEMIN8
Genomic Map