Assay Details
| Gene Symbol: FH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075803.2 | NP_001069271.1 | 4 - 5 | 579 | 70 | |
| GenBank mRNA | BC118450.1 | - | 4 - 5 | 580 | 70 | |
More Information
Set Membership: |
|

FH
Genomic Map