Assay Details
| Gene Symbol: SDR39U1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075812.2 | NP_001069280.1 | 3 - 4 | 280 | 72 | |
| GenBank mRNA | BC118349.1 | - | 3 - 4 | 282 | 72 | |
More Information
Set Membership: |
|

SDR39U1
Genomic Map