Assay Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075879.1 | NP_001069347.1 | 2 - 3 | 369 | 113 | |
| GenBank mRNA | BC116145.1 | - | 2 - 3 | 369 | 113 | |
More Information
Set Membership: |
|

CHST9
Genomic Map