Assay Details
| Gene Symbol: SLC52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075901.2 | NP_001069369.1 | 3 - 4 | 1535 | 69 | |
| GenBank mRNA | BC118403.1 | - | 3 - 4 | 1536 | 69 | |
More Information
Set Membership: |
|

SLC52A2
Genomic Map