Assay Details
| Gene Symbol: LRRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075904.1 | NP_001069372.1 | 9 - 10 | 1133 | 61 | |
| GenBank mRNA | BC114759.1 | - | 9 - 10 | 1133 | 61 | |
More Information
Set Membership: |
|

LRRC6
Genomic Map