Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075941.2 | NP_001069409.2 | 1 - 2 | 225 | 150 | |
| GenBank mRNA | BC118142.1 | - | 1 - 2 | 185 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118142.1, NM_001075941.2 transcripts.
Set Membership: |
|

CLN8
Genomic Map