Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075941.2 | NP_001069409.2 | 2 - 3 | 295 | 112 | |
| GenBank mRNA | BC118142.1 | - | 2 - 3 | 255 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118142.1, NM_001075941.2 transcripts.
Set Membership: |
|

CLN8
Genomic Map