Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076045.2 | NP_001069513.1 | 2 - 3 | 348 | 88 | |
| GenBank mRNA | BC116149.1 | - | 2 - 3 | 349 | 88 | |
More Information
Set Membership: |
|

LACTB2
Genomic Map