Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076071.2 | NP_001069539.1 | 6 - 7 | 1378 | 64 | |
| GenBank mRNA | BC114021.1 | - | 6 - 7 | 1379 | 64 | |
More Information
Set Membership: |
|

RNF128
Genomic Map