Assay Details
| Gene Symbol: RHOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076099.1 | NP_001069567.1 | 1 - 2 | 284 | 120 | |
| GenBank mRNA | BC118335.1 | - | 1 - 2 | 284 | 120 | |
More Information
Set Membership: |
|

RHOV
Genomic Map