Assay Details
| Gene Symbol: POLR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076144.2 | NP_001069612.1 | 2 | 484 | 91 | |
| GenBank mRNA | BC114905.1 | - | 1 | 448 | 91 | |
| DT857457.1 | - | 1 | 91 | |||
| DT857482.1 | - | 1 | 478 | 91 | ||
More Information
Set Membership: |
|

POLR1D
Genomic Map