Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076154.2 | NP_001069622.1 | 2 - 3 | 334 | 87 | |
| GenBank mRNA | BC118390.1 | - | 2 - 3 | 336 | 87 | |
More Information
Set Membership: |
|

CFL2
Genomic Map