Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076303.2 | NP_001069771.1 | 3 - 4 | 315 | 65 | |
| GenBank mRNA | BC118358.1 | - | 3 - 4 | 316 | 65 | |
More Information
Set Membership: |
|

COLEC11
Genomic Map