Assay Details
| Gene Symbol: NUDT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076314.1 | NP_001069782.1 | 6 - 7 | 581 | 60 | |
| GenBank mRNA | BC118134.1 | - | 6 - 7 | 581 | 60 | |
More Information
Set Membership: |
|

NUDT5
Genomic Map