Assay Details
| Gene Symbol: C7H19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076345.1 | NP_001069813.1 | 1 - 2 | 106 | 96 | |
| GenBank mRNA | BC118308.1 | - | 1 - 2 | 106 | 96 | |
More Information
Set Membership: |
|

C7H19orf53
Genomic Map