Assay Details
| Gene Symbol: TIGAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076370.1 | NP_001069838.1 | 2 - 3 | 109 | 76 | |
| GenBank mRNA | BC116101.1 | - | 2 - 3 | 109 | 76 | |
More Information
Set Membership: |
|

TIGAR
Genomic Map