Assay Details
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076453.2 | NP_001069921.1 | 4 - 5 | 419 | 55 | |
| GenBank mRNA | BC118431.1 | - | 4 - 5 | 420 | 55 | |
More Information
Set Membership: |
|

SLC19A1
Genomic Map