Assay Details
| Gene Symbol: COA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076459.2 | NP_001069927.1 | 4 - 5 | 498 | 106 | |
| GenBank mRNA | BC118424.1 | - | 4 - 5 | 500 | 106 | |
| BT025435.1 | - | 4 - 5 | 472 | 106 | ||
More Information
Set Membership: |
|

COA8
Genomic Map