Assay Details
| Gene Symbol: UBE2S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076472.2 | NP_001069940.1 | 2 - 3 | 387 | 75 | |
| GenBank mRNA | BC114838.1 | - | 2 - 3 | 388 | 75 | |
| BC148138.1 | - | 2 - 3 | 350 | 75 | ||
More Information
Set Membership: |
|

UBE2S
Genomic Map