Assay Details
| Gene Symbol: GEMIN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076489.1 | NP_001069957.1 | 1 - 2 | 39 | 58 | |
| GenBank mRNA | BC118472.1 | - | 1 - 2 | 39 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118472.1, NM_001076489.1 transcripts.
Set Membership: |
|

GEMIN7
Genomic Map