Assay Details
| Gene Symbol: SULT4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076518.1 | NP_001069986.1 | 4 - 5 | 563 | 62 | |
| GenBank mRNA | BC118153.1 | - | 4 - 5 | 563 | 62 | |
More Information
Set Membership: |
|

SULT4A1
Genomic Map