Assay Details
| Gene Symbol: FBLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076955.1 | NP_001070423.1 | 4 - 5 | 589 | 126 | |
| GenBank mRNA | BC116079.1 | - | 4 - 5 | 589 | 126 | |
More Information
Set Membership: |
|

FBLIM1
Genomic Map