Assay Details
| Gene Symbol: C8H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076978.1 | NP_001070446.1 | 1 - 2 | 575 | 70 | |
| GenBank mRNA | BT025481.1 | - | 1 - 2 | 575 | 70 | |
More Information
Set Membership: |
|

C8H9orf64
Genomic Map