Assay Details
| Gene Symbol: C8H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076978.1 | NP_001070446.1 | 2 - 3 | 833 | 104 | |
| GenBank mRNA | BT025481.1 | - | 2 - 3 | 833 | 104 | |
More Information
Set Membership: |
|

C8H9orf64
Genomic Map