Assay Details
| Gene Symbol: AP1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077110.1 | NP_001070578.1 | 1 - 2 | 685 | 63 | |
| GenBank mRNA | BC116132.1 | - | 1 - 2 | 685 | 63 | |
More Information
Set Membership: |
|

AP1S1
Genomic Map