Assay Details
| Gene Symbol: GSTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077112.1 | NP_001070580.1 | 5 - 6 | 461 | 91 | |
| GenBank mRNA | BC116161.1 | - | 5 - 6 | 461 | 91 | |
More Information
Set Membership: |
|

GSTA3
Genomic Map