Assay Details
| Gene Symbol: ESAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078066.2 | NP_001071534.1 | 4 - 5 | 770 | 61 | |
| GenBank mRNA | BC105434.1 | - | 4 - 5 | 771 | 61 | |
More Information
Set Membership: |
|

ESAM
Genomic Map