Assay Details
| Gene Symbol: HSP90AB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079637.1 | NP_001073105.1 | 11 - 12 | 2155 | 60 | |
| GenBank mRNA | AB072369.1 | - | 11 - 12 | 2150 | 60 | |
| BT025368.1 | - | 11 - 12 | 2155 | 60 | ||
More Information
Set Membership: |
|

HSP90AB1
Genomic Map