Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080225.1 | NP_001073694.1 | 2 - 3 | 362 | 64 | |
| GenBank mRNA | BC118191.1 | - | 2 - 3 | 362 | 64 | |
More Information
Set Membership: |
|

LRPAP1
Genomic Map