Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080231.1 | NP_001073700.2 | 1 - 2 | 879 | 105 | |
| NM_001244294.1 | NP_001231223.1 | 2 - 3 | 296 | 105 | ||
| GenBank mRNA | BC116008.1 | - | 1 - 2 | 879 | 105 | |
| BC151424.1 | - | 2 - 3 | 216 | 105 | ||
| DN542857.1 | - | 2 - 3 | 296 | 105 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map