Assay Details
| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080349.2 | NP_001073818.1 | 2 - 3 | 262 | 68 | |
| GenBank mRNA | BC118400.1 | - | 2 - 3 | 263 | 68 | |
More Information
Set Membership: |
|

EEF1AKMT1
Genomic Map