Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174195.3 | NP_776620.2 | 7 - 8 | 1573 | 73 | |
| GenBank mRNA | AF121289.1 | - | 7 - 8 | 1120 | 73 | |
| BC114678.1 | - | 7 - 8 | 1573 | 73 | ||
| BT021579.1 | - | 7 - 8 | 1300 | 73 | ||
| BT021659.1 | - | 7 - 8 | 1206 | 73 | ||
| BT021856.1 | - | 7 - 8 | 1155 | 73 | ||
| BT021880.1 | - | 7 - 8 | 1227 | 73 | ||
| BT021899.1 | - | 7 - 8 | 1304 | 73 | ||
| BT026178.1 | - | 7 - 8 | 1169 | 73 | ||
| BT026282.1 | - | 7 - 8 | 1206 | 73 | ||
More Information
Set Membership: |
|

TCN2
Genomic Map