Assay Details
| Gene Symbol: CASP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176638.5 | NP_788811.1 | 4 - 5 | 569 | 74 | |
| GenBank mRNA | AF383946.1 | - | 4 - 5 | 556 | 74 | |
| AF383947.1 | - | 3 - 4 | 385 | 74 | ||
| BC112708.1 | - | 4 - 5 | 570 | 74 | ||
| BT020999.1 | - | 4 - 5 | 586 | 74 | ||
More Information
Set Membership: |
|

CASP4
Genomic Map