Assay Details
| Gene Symbol: FOXRED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034371.3 | NP_001029543.2 | 5 - 6 | 732 | 59 | |
| GenBank mRNA | BC120036.1 | - | 5 - 6 | 733 | 59 | |
| BT020666.1 | - | 5 - 6 | 678 | 59 | ||
| BT020724.1 | - | 5 - 6 | 679 | 59 | ||
| BT021902.1 | - | 5 - 6 | 681 | 59 | ||
More Information
Set Membership: |
|

FOXRED1
Genomic Map