Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034380.2 | NP_001029552.2 | 3 - 4 | 384 | 108 | |
| GenBank mRNA | BC102717.1 | - | 3 - 4 | 210 | 108 | |
| BT026308.1 | - | 3 - 4 | 384 | 108 | ||
More Information
Set Membership: |
|

CFAP126
Genomic Map