Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC120245.1 | - | 1 - 2 | 44 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120245.1 transcripts.
Set Membership: |
|

GPR85
Genomic Map